Learn More
Dwyer Instruments Inc 1/4"OD QUICK COUPLING X1/4 NPT
Supplier: Dwyer Instruments Inc A30212
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
1/4" O.D. quick coupling x 1/4" male NPT straight metal fitting.
This item is not returnable.
View return policy