Learn More
SKYLINE CENTER Item #W85 Bag 1 3/4” Square - 500ct.
Supplier: SKYLINE CENTER W85GUN PATCH
1 3/4” SQUARE 100% COTTON PATCHES Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment. Gun Cleaning - 7mm - .38/.357 Caliber