Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane (Spectranalyzed™), Fisher Chemical

Click to view available options
Quantity:
4 L
Packaging:
Amber Glass
This item is not returnable. View return policy
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
11
CHEBI:27789
1,2-dichloroethane
ClCCCl
This item is not returnable. View return policy
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
11
CHEBI:27789
1,2-dichloroethane
ClCCCl
Melting Point -35°C
Absorbance Pass Test
Color Colorless
Assay Percent Range ≥99 %
Formula Weight 98.96
Percent Purity ≥99%
Grade Also meets ACS specifications
Density 1.250g/cm³
Boiling Point 81°C
Quantity 4 L
Identification Pass Test
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation 0.002% max.
Titratable Acid 0.0003mEq/g max.
Vapor Pressure 65mmHg at 29°C
Viscosity 0.8 mPa/s at 20°C
Water 0.03% max.
Chemical Name or Material 1, 2-Dichloroethane
Show More Show Less

DANGER!

Emergency Overview
Flammable liquid and vapor. Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. May cause adverse liver effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit