Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethane, Anhydrous, Approx. 99.8%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
ClCCCl
This item is not returnable. View return policy
107-06-2
C2H4Cl2
98.95
MFCD00000963
WSLDOOZREJYCGB-UHFFFAOYSA-N
1,2-dichloroethane
ClCCCl

D1386, 107-06-2, C2H4Cl2

Spectrum™ Chemical 1,2-Dichloroethane, Anhydrous is among Spectrum Chemicals′s high quality inventory of research chemicals. Also known as Ethylene Dichloride or Ethylene dichloride, 1,2-Dichloroethane is a chemical compound and chlorinated hydrocarbon. It is a colorless liquid with a chloroform-like scent and sweet taste, and is most often used in the production of vinyl chloride. Anhydrous solvents are used when performing certain reactions where the presence of water can prevent a chemical reaction from happening or form undesirable products. Spectrum anhydrous chemical solvents are packaged in convenient sizes featuring a double closure, which includes an easy-slip-on vinyl cap with Teflon liner and screw-on phenolic cap. These items are packaged in specially designed septum-sealed bottles under nitrogen to maintain low water content and ensure product integrity.
CAS Max % 100%
UN Number UN1184
Percent Purity Approx. 99.8%
Grade Reagent
Quantity 1 L
Packaging Amber Glass Bottle
Residue after Evaporation 0.03%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit