Learn More
Cole-Parmer Hydrion 165/1-12 pH Paper Strips, Range 1-12, 100 Strips/Vial, 10 Vials/Carton
Supplier: Cole-Parmer 5920025
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These low-cost disposable vials of pHydrion pH paper strips test pH ranging from 0 to 12. Box of ten vials; 100 strips per vial.