Learn More
Nelson Jameson Company 3M Petrifilm E.coli/Coliform Count Plates, 50/pk
Supplier: Nelson Jameson Company 6404
E.coli/Coliform Count Plates identify both E.coli and other coliforms. With this one easy test, you'll have confirmed results in 24 to 48 hours. Maximize productivity by increasing your laboratory efficiency with 3M Petrifilm Plates.