Learn More
Ace Glass, Inc. -008 O-RING FETFE 12/PK -008 O-RING FETFE 12/PK
Supplier: Ace Glass, Inc. 7855704
Size 008 FETFE o-ring, Pack/12. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.