Learn More
Fine Science Tools 0.5MM DIA DRILL BURR PK/100.5
SDP
Supplier:  Fine Science Tools 1900705
Encompass
Tip Diameter: 0.5mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Carbon Steel/ Length: 44mm/ Autoclave Safe: No
Catalog No. NC9152343
Explore available promotions