Learn More
Cole-Parmer Cole-Parmer +/-0.25%-Accuracy Transmitter, 0 to 1 psi; 4 to 20 mA Output
Supplier: Cole-Parmer 6807501
Economical, excellent accuracy, and high performance. Ideal for industrial OEM equipment, engines, hydraulic systems, and compressor control.
Transducers with 4 to 20 mA output have red (positive power) and black (output) wires. Transducers with 0.5 to 5.5 V output have red (positive power), green (output), and black/white (common) wires.