Assay Details
Test
Viewing profile as user:
Sign In
Don't have an account?
Create an Account
Account
Main Menu
Admin
Main Menu
Admin
ULS Inventory Manager
Main Menu
ULS Inventory Manager
Products
Antibodies
Products
Antibodies
Chemicals
Consumables
Equipment and Instruments
Furniture and Storage
Life Sciences Products
Products
Life Sciences Products
Safety Products
Products
Safety Products
Applications
Tools
Assay Tools
Chemical Structure Tools
Tools
Chemical Structure Tools
Oligos Tools
Tools
Programs and Services
Services
Programs and Services
Documents and Certificates
Main Menu
Special Offers
Main Menu
Special Offers
Orders
Main Menu
Shopping Tools
Main Menu
Please try again - we encountered an error while attempting to complete your request.
Contact website technical assistance if you see this message again.
Your items have has been added!
Host server : magellan-srch-1-prod-blue:8080/10.253.225.184:8080.
git-commit: 9c2c959b77162a04e431fec2cb3661656af8b157
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.4.0-2024.02.08-1.0
git-commit: 9c2c959b77162a04e431fec2cb3661656af8b157
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.4.0-2024.02.08-1.0