Assay Details
Mature miRNA Sequence: | GUGUGGAAGAUGGGAGGAGAAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6833-5p | MIMAT0027566 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6833-5p | ||
---|---|---|---|
miRBase Accession Number: | MI0022678 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.6: 32179816 - 32179876 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | GUGUGGAAGAUGGGAGGAGAAA | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | GUGUGGAAGAUGGGAGGAGAAA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6833-5p | MIMAT0027566 |
Stem-loop ID | hsa-mir-6833 |
Stem-loop Accession # | MI0022678 |
Stem-loop Sequence | AAACGGUGUGGAAGAUGGGAGGAGAAAAAUCCCUGUUAACUUUCUCUCUCCACUUCCUCAG |
Chromosome Location | Chr. 6 - 32179816 - 32179876 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027566
miRBase ID: hsa-miR-6833-5p Mature miRNA Sequence: GUGUGGAAGAUGGGAGGAGAAA Chromosome Location: Chr. 6 - 32179816 - 32179876 [+] on Build GRCh38 |