Assay Details
Gene Family ID: | MIPF0001488, mir-3188 |
Mature miRNA Sequence: | AGAGGCUUUGUGCGGAUACGGGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3188 | MIMAT0015070 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3188 | ||
---|---|---|---|
miRBase Accession Number: | MI0014232 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.19: 18282077 - 18282161 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AGAGGCUUUGUGCGGAUACGGGG | ||
Species: | Human | ||
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001488, mir-3188 |
Mature miRNA Sequence: | AGAGGCUUUGUGCGGAUACGGGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3188 | MIMAT0015070 |
Stem-loop ID | hsa-mir-3188 |
Stem-loop Accession # | MI0014232 |
Stem-loop Sequence | GGCGCCUCCUGCUCUGCUGUGCCGCCAGGGCCUCCCCUAGCGCGCCUUCUGGAGAGGCUUUGUGCGGAUACGGGGCUGGAGGCCU |
Chromosome Location | Chr. 19 - 18282077 - 18282161 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0015070
miRBase ID: hsa-miR-3188 Mature miRNA Sequence: AGAGGCUUUGUGCGGAUACGGGG Chromosome Location: Chr. 19 - 18282077 - 18282161 [+] on Build GRCh38 |