Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CAAAGACUGCAAUUACUUUUGCG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548u | MIMAT0015013 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-548u | ||
---|---|---|---|
miRBase Accession Number: | MI0014168 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.6: 57390132 - 57390212 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | CAAAGACUGCAAUUACUUUUGCG | ||
Species: | Human | ||
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CAAAGACUGCAAUUACUUUUGCG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548u | MIMAT0015013 |
Stem-loop ID | hsa-mir-548u |
Stem-loop Accession # | MI0014168 |
Stem-loop Sequence | AUUAGGAUGGUGCAAAAGUAAUGUGGUUUUUUUCUUUACUUUUAAUGGCAAAGACUGCAAUUACUUUUGCGCCAACCUAAU |
Chromosome Location | Chr. 6 - 57390132 - 57390212 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0015013
miRBase ID: hsa-miR-548u Mature miRNA Sequence: CAAAGACUGCAAUUACUUUUGCG Chromosome Location: Chr. 6 - 57390132 - 57390212 [+] on Build GRCh38 |