Assay Details
Gene Family ID: | MIPF0000148, mir-451 |
Mature miRNA Sequence: | AAACCGUUACCAUUACUGAGUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-451a | MIMAT0001631 |
Mouse | mmu-miR-451a | MIMAT0001632 |
Rat | rno-miR-451-5p | MIMAT0001633 |
Anolis carolinensis | aca-miR-451-5p | MIMAT0021960 |
Cricetulus griseus | cgr-miR-451a | MIMAT0041141 |
Dog | cfa-miR-451 | MIMAT0009870 |
View More Rows | ||
Pan troglodytes | ptr-miR-451 | MIMAT0008153 |
Pig | ssc-miR-451 | MIMAT0018382 |
Pongo pygmaeus | ppy-miR-451 | MIMAT0015896 |
Rhesus monkey | mml-miR-451 | MIMAT0006335 |
Xenopus tropicalis | xtr-miR-451 | MIMAT0003705 |
Zebrafish | dre-miR-451 | MIMAT0001634 |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Cricetulus griseus
Dog
Pan troglodytes
Pig
Pongo pygmaeus
Rhesus monkey
Xenopus tropicalis
Zebrafish
More Information
Related Products
miRBase Alias:
Human: hsa-miR-451 (v17)
Rat: rno-miR-451 (v18)
Aeromonas caviae Ae398: aca-miR-451 (v18)
Macaca mulatta polyomavirus 1: mmu-miR-451 (v18)
Rat: rno-miR-451 (v18)
Aeromonas caviae Ae398: aca-miR-451 (v18)
Macaca mulatta polyomavirus 1: mmu-miR-451 (v18)