Assay Details
Mature miRNA Sequence: | AGAAUUCUCUUAUCCAACAUCAACA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-10393-5p | MIMAT0041617 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-10393-5p | ||
---|---|---|---|
miRBase Accession Number: | MI0033417 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.15: 44717844 - 44717893 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AGAAUUCUCUUAUCCAACAUCAACA | ||
Species: | Human | ||
Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | AGAAUUCUCUUAUCCAACAUCAACA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-10393-5p | MIMAT0041617 |
Stem-loop ID | hsa-mir-10393 |
Stem-loop Accession # | MI0033417 |
Stem-loop Sequence | AGAAUUCUCUUAUCCAACAUCAACAUCUUGGUCAGAUUUGAACUCUUCAA |
Chromosome Location | Chr. 15 - 44717844 - 44717893 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0041617
miRBase ID: hsa-miR-10393-5p Mature miRNA Sequence: AGAAUUCUCUUAUCCAACAUCAACA Chromosome Location: Chr. 15 - 44717844 - 44717893 [+] on Build GRCh38 |