Assay Details
Gene Family ID: | MIPF0001623, mir-4488 |
Mature miRNA Sequence: | AGGGGGCGGGCUCCGGCG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4488 | MIMAT0019022 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4488 | ||
---|---|---|---|
miRBase Accession Number: | MI0016849 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.11: 61508596 - 61508657 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AGGGGGCGGGCUCCGGCG | ||
Species: | Human | ||
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0001623, mir-4488 |
Mature miRNA Sequence: | AGGGGGCGGGCUCCGGCG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4488 | MIMAT0019022 |
Stem-loop ID | hsa-mir-4488 |
Stem-loop Accession # | MI0016849 |
Stem-loop Sequence | GGUAGGGGGCGGGCUCCGGCGCUGGGACCCCACUAGGGUGGCGCCUUGGCCCCGCCCCGCCC |
Chromosome Location | Chr. 11 - 61508596 - 61508657 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019022
miRBase ID: hsa-miR-4488 Mature miRNA Sequence: AGGGGGCGGGCUCCGGCG Chromosome Location: Chr. 11 - 61508596 - 61508657 [+] on Build GRCh38 |