Assay Details
Gene Family ID: | MIPF0000148, mir-451 |
Mature miRNA Sequence: | UAGCAAGAGAACCAUUACCAUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-451b | MIMAT0019840 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-451b | ||
---|---|---|---|
miRBase Accession Number: | MI0017360 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.17: 28861371 - 28861438 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UAGCAAGAGAACCAUUACCAUU | ||
Species: | Human | ||
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000148, mir-451 |
Mature miRNA Sequence: | UAGCAAGAGAACCAUUACCAUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-451b | MIMAT0019840 |
Stem-loop ID | hsa-mir-451b |
Stem-loop Accession # | MI0017360 |
Stem-loop Sequence | UGGGUAUAGCAAGAGAACCAUUACCAUUACUAAACUCAGUAAUGGUAACGGUUUCCUUGCCAUUCCCA |
Chromosome Location | Chr. 17 - 28861371 - 28861438 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019840
miRBase ID: hsa-miR-451b Mature miRNA Sequence: UAGCAAGAGAACCAUUACCAUU Chromosome Location: Chr. 17 - 28861371 - 28861438 [+] on Build GRCh38 |