Assay Details
Amplicon Length: | 135 |
Chromosome Location: | Chr. 12 - 57512505 - 57512505 [-] on 57512505 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6758-3p | MIMAT0027417 |
Human | hsa-miR-6758-5p | MIMAT0027416 |
Stem-loop Details
Human