Assay Details
Amplicon Length: | 78 |
Chromosome Location: | Chr. 21 - 45478490 - 45478490 [+] on 45478490 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6815-3p | MIMAT0027531 |
Human | hsa-miR-6815-5p | MIMAT0027530 |
Stem-loop Details
Human