Assay Details
Amplicon Length: | 90 |
Chromosome Location: | Chr. 1 - 37480450 - 37480450 [-] on 37480450 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6732-5p | MIMAT0027365 |
Human | hsa-miR-6732-3p | MIMAT0027366 |
Stem-loop Details
Human