Assay Details
Amplicon Length: | 76 |
Chromosome Location: | Chr. 19 - 8388909 - 8388909 [+] on 8388909 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4999-5p | MIMAT0021017 |
Human | hsa-miR-4999-3p | MIMAT0021018 |
Stem-loop Details
Human