Assay Details
Amplicon Length: | 104 |
Chromosome Location: | Chr. X - 47587274 - 47587274 [+] on 47587274 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4769-5p | MIMAT0019922 |
Human | hsa-miR-4769-3p | MIMAT0019923 |
Stem-loop Details
Human