Assay Details
Amplicon Length: | 90 |
Chromosome Location: | Chr. 6 - 41598568 - 41598568 [+] on 41598568 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4641 | MIMAT0019701 |
Stem-loop Details
Human
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)