Assay Details
Amplicon Length: | 92 |
Chromosome Location: | Chr. 22 - 30731806 - 30731806 [-] on 30731806 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3200-3p | MIMAT0015085 |
Human | hsa-miR-3200-5p | MIMAT0017392 |
Stem-loop Details
Human