Assay Details
Amplicon Length: | 143 |
Chromosome Location: | Chr. 3 - 31161552 - 31161552 [-] on 31161552 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-466 | MIMAT0015002 |
Stem-loop Details
Human