Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. 12 - 97995236 - 97995236 [+] on 97995236 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4303 | MIMAT0016856 |
Stem-loop Details
Human