Assay Details
Amplicon Length: | 71 |
Chromosome Location: | Chr. 17 - 75405924 - 75405924 [-] on 75405924 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3678-5p | MIMAT0018102 |
Human | hsa-miR-3678-3p | MIMAT0018103 |
Stem-loop Details
Human