Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 14 - 101066526 - 101066526 [+] on 101066526 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-656-3p | MIMAT0003332 |
Human | hsa-miR-656-5p | MIMAT0026627 |
Stem-loop Details
Human