Assay Details
Amplicon Length: | 79 |
Chromosome Location: | Chr. 10 - 100974687 - 100974687 [+] on 100974687 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-608 | MIMAT0003276 |
Stem-loop Details
Human