Assay Details
Gene Family ID: | MIPF0000546, mir-633 |
Mature miRNA Sequence: | CUAAUAGUAUCUACCACAAUAAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-633 | MIMAT0003303 |
Rhesus monkey | mml-miR-633 | MIMAT0006478 |
Stem-loop Details
Human
Rhesus monkey