Assay Details
Gene Family ID: | MIPF0000465, mir-661 |
Mature miRNA Sequence: | UGCCUGGGUCUCUGGCCUGCGCGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-661 | MIMAT0003324 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-661 | ||
---|---|---|---|
miRBase Accession Number: | MI0003669 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.8: 143945191 - 143945279 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UGCCUGGGUCUCUGGCCUGCGCGU | ||
Species: | Human | ||
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000465, mir-661 |
Mature miRNA Sequence: | UGCCUGGGUCUCUGGCCUGCGCGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-661 | MIMAT0003324 |
Stem-loop ID | hsa-mir-661 |
Stem-loop Accession # | MI0003669 |
Stem-loop Sequence | GGAGAGGCUGUGCUGUGGGGCAGGCGCAGGCCUGAGCCCUGGUUUCGGGCUGCCUGGGUCUCUGGCCUGCGCGUGACUUUGGGGUGGCU |
Chromosome Location | Chr. 8 - 143945191 - 143945279 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0003324
miRBase ID: hsa-miR-661 Mature miRNA Sequence: UGCCUGGGUCUCUGGCCUGCGCGU Chromosome Location: Chr. 8 - 143945191 - 143945279 [-] on Build GRCh38 |