Assay Details
Gene Family ID: | MIPF0000046, mir-101 |
Mature miRNA Sequence: | UACAGUACUGUGAUAACUGAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-101-3p | MIMAT0000099 |
Mouse | mmu-miR-101a-3p | MIMAT0000133 |
Rat | rno-miR-101a-3p | MIMAT0000823 |
Bovine | bta-miR-101 | MIMAT0003520 |
Callithrix jacchus | cja-miR-101-2 | MIMAT0049331 |
Columba livia | cli-miR-101-3p | MIMAT0038483 |
View More Rows | ||
Horse | eca-miR-101 | MIMAT0012951 |
Pig | ssc-miR-101 | MIMAT0010185 |
Pongo pygmaeus | ppy-miR-101 | MIMAT0002429 |
Stem-loop Details
Human
Stem-loop ID | hsa-mir-101-1 |
Stem-loop Accession # | MI0000103 |
Stem-loop Sequence | UGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCUAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCA |
Chromosome Location | Chr. 1 - 65058434 - 65058508 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000099
miRBase ID: hsa-miR-101-3p Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: Chr. 1 - 65058434 - 65058508 [-] on Build GRCh38 |
Stem-loop ID | hsa-mir-101-2 |
Stem-loop Accession # | MI0000739 |
Stem-loop Sequence | ACUGUCCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGU |
Chromosome Location | Chr. 9 - 4850297 - 4850375 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000099
miRBase ID: hsa-miR-101-3p Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: Chr. 1 - 65058434 - 65058508 [-] on Build GRCh38 |
Stem-loop ID | mmu-mir-101a |
Stem-loop Accession # | MI0000148 |
Stem-loop Sequence | AGGCUGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCCAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCAGCCA |
Chromosome Location | Chr. 4 - 101346945 - 101347027 [-] on Build GRCm38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000133
miRBase ID: mmu-miR-101a-3p Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: Chr. 4 - 101346945 - 101347027 [-] on Build GRCm38 |
Stem-loop ID | rno-mir-101a |
Stem-loop Accession # | MI0000886 |
Stem-loop Sequence | UGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCCAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCA |
Chromosome Location | Chr. 5 - 120168437 - 120168511 [-] on Build Rnor_6.0 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000823
miRBase ID: rno-miR-101a-3p Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: Chr. 5 - 120168437 - 120168511 [-] on Build Rnor_6.0 |
Stem-loop ID | bta-mir-101-1 |
Stem-loop Accession # | MI0009721 |
Stem-loop Sequence | AGGCUGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCCAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCAGCCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003520
miRBase ID: bta-miR-101 Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
Stem-loop ID | bta-mir-101-2 |
Stem-loop Accession # | MI0004735 |
Stem-loop Sequence | ACUGUCCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003520
miRBase ID: bta-miR-101 Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
Stem-loop ID | cja-mir-101-2 |
Stem-loop Accession # | MI0040067 |
Stem-loop Sequence | CGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049331
miRBase ID: cja-miR-101-2 Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
Stem-loop ID | cli-mir-101-1 |
Stem-loop Accession # | MI0029947 |
Stem-loop Sequence | ACAGGCUGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCUCUUGUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCAGCCAUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038483
miRBase ID: cli-miR-101-3p Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
Stem-loop ID | cli-mir-101-2 |
Stem-loop Accession # | MI0029948 |
Stem-loop Sequence | ACUAUGAACUGUCCUUUUUCGGUUAUCAUGGUACCGGUGCUGUAUACGUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGUGCCAUCAC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038483
miRBase ID: cli-miR-101-3p Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
Stem-loop ID | eca-mir-101-1 |
Stem-loop Accession # | MI0012703 |
Stem-loop Sequence | CUCAGUUAUCACAGUGCUGAUGCUGUCAGUUCUAAAGGUACAGUACUGUGAUAACUGAA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0012951
miRBase ID: eca-miR-101 Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
Stem-loop ID | eca-mir-101-2 |
Stem-loop Accession # | MI0012861 |
Stem-loop Sequence | CCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0012951
miRBase ID: eca-miR-101 Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
Stem-loop ID | ssc-mir-101-1 |
Stem-loop Accession # | MI0010678 |
Stem-loop Sequence | GACUGAACUGUCCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGUGCCAUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0010185
miRBase ID: ssc-miR-101 Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
Stem-loop ID | ssc-mir-101-2 |
Stem-loop Accession # | MI0010679 |
Stem-loop Sequence | GGCUGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCCAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCAGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0010185
miRBase ID: ssc-miR-101 Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
Stem-loop ID | ppy-mir-101-1 |
Stem-loop Accession # | MI0014811 |
Stem-loop Sequence | UGCCCUGGCUCAGUUAUCACAGUGCUGAUGCUGUCCAUUCUAAAGGUACAGUACUGUGAUAACUGAAGGAUGGCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002429
miRBase ID: ppy-miR-101 Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
Stem-loop ID | ppy-mir-101-2 |
Stem-loop Accession # | MI0014812 |
Stem-loop Sequence | ACUGUCCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002429
miRBase ID: ppy-miR-101 Mature miRNA Sequence: UACAGUACUGUGAUAACUGAA Chromosome Location: NA |
More Information
Related Products
TaqMan™ Pri-miRNA Assay : Mm03306215_pri
TaqMan™ Pri-miRNA Assay : Rn03464794_pri
TaqMan™ Pri-miRNA Assay : Hs03302741_pri
TaqMan™ Pri-miRNA Assay : Hs03303387_pri
Ambion® Pre-miR™ miRNA Precursor : PM11414
TaqMan™ MicroRNA Assay : 002253
mirVana® miRNA inhibitor : MH11414
mirVana® miRNA mimic : MC11414
TaqMan™ Advanced miRNA Assay : 477863_mir
TaqMan™ Advanced miRNA Assay : mmu483018_mir
TaqMan™ Pri-miRNA Assay : Rn03464794_pri
TaqMan™ Pri-miRNA Assay : Hs03302741_pri
TaqMan™ Pri-miRNA Assay : Hs03303387_pri
Ambion® Pre-miR™ miRNA Precursor : PM11414
TaqMan™ MicroRNA Assay : 002253
mirVana® miRNA inhibitor : MH11414
mirVana® miRNA mimic : MC11414
TaqMan™ Advanced miRNA Assay : 477863_mir
TaqMan™ Advanced miRNA Assay : mmu483018_mir
miRBase Alias:
Human: hsa-miR-101 (v17)
Rat: rno-miR-101a (v18)
Macaca mulatta polyomavirus 1: mmu-miR-101a (v17)
Staphylococcus schleiferi: ssc-miR-101a (v14)
Rat: rno-miR-101a (v18)
Macaca mulatta polyomavirus 1: mmu-miR-101a (v17)
Staphylococcus schleiferi: ssc-miR-101a (v14)