Assay Details
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | AACCCGUAGAUCCGAUCUUGUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-99a-5p | MIMAT0000097 |
Mouse | mmu-miR-99a-5p | MIMAT0000131 |
Rat | rno-miR-99a-5p | MIMAT0000820 |
Alligator mississippiensis | ami-miR-99a-5p | MIMAT0038110 |
Anolis carolinensis | aca-miR-99a-5p | MIMAT0021996 |
Chicken | gga-miR-99a-5p | MIMAT0001103 |
View More Rows | ||
Chrysemys picta | cpi-miR-99a-5p | MIMAT0037710 |
Columba livia | cli-miR-99-5p | MIMAT0038478 |
Dasypus novemcinctus | dno-miR-99a-5p | MIMAT0047594 |
Gadus morhua | gmo-miR-99-5p | MIMAT0044269 |
Gorilla gorilla | ggo-miR-99a | MIMAT0002412 |
Guinea pig | cpo-miR-99a-5p | MIMAT0046941 |
Horse | eca-miR-99a | MIMAT0013184 |
Ictalurus punctatus | ipu-miR-99a | MIMAT0029532 |
Lagothrix lagotricha | lla-miR-99a | MIMAT0002414 |
Macaca nemestrina | mne-miR-99a | MIMAT0002415 |
Metriaclima zebra | mze-miR-99b | MIMAT0042328 |
Microcebus murinus | mmr-miR-99a | MIMAT0049202 |
Neolamprologus brichardi | nbr-miR-99b | MIMAT0042505 |
Ophiophagus hannah | oha-miR-99a-5p | MIMAT0036964 |
Oreochromis niloticus | oni-miR-99b | MIMAT0042695 |
Pan paniscus | ppa-miR-99a | MIMAT0002416 |
Pan troglodytes | ptr-miR-99a | MIMAT0002411 |
Pig | ssc-miR-99a-5p | MIMAT0013896 |
Pongo pygmaeus | ppy-miR-99a | MIMAT0002413 |
Pundamilia nyererei | pny-miR-99b | MIMAT0042888 |
Rabbit | ocu-miR-99a-5p | MIMAT0048182 |
Rhesus monkey | mml-miR-99a-5p | MIMAT0002410 |
Salmo salar | ssa-miR-99-5p | MIMAT0032718 |
Tupaia chinensis | tch-miR-99a-5p | MIMAT0036563 |
Xenopus tropicalis | xtr-miR-99 | MIMAT0003582 |
Zebrafish | dre-miR-99 | MIMAT0001812 |
Stem-loop Details
Human
Stem-loop ID | hsa-mir-99a |
Stem-loop Accession # | MI0000101 |
Stem-loop Sequence | CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG |
Chromosome Location | Chr. 21 - 16539089 - 16539169 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000097
miRBase ID: hsa-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: Chr. 21 - 16539089 - 16539169 [+] on Build GRCh38 |
Stem-loop ID | mmu-mir-99a |
Stem-loop Accession # | MI0000146 |
Stem-loop Sequence | CAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCGCAAGCUCGUUUCUAUGGGUCUGUG |
Chromosome Location | Chr. 16 - 77598936 - 77599000 [+] on Build GRCm38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000131
miRBase ID: mmu-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: Chr. 16 - 77598936 - 77599000 [+] on Build GRCm38 |
Stem-loop ID | rno-mir-99a |
Stem-loop Accession # | MI0000883 |
Stem-loop Sequence | CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGUUUCUAUGGGUCUGUGGCAGUGUG |
Chromosome Location | Chr. 11 - 16052153 - 16052233 [+] on Build Rnor_6.0 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000820
miRBase ID: rno-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: Chr. 11 - 16052153 - 16052233 [+] on Build Rnor_6.0 |
Stem-loop ID | ami-mir-99a |
Stem-loop Accession # | MI0029708 |
Stem-loop Sequence | GUGCCAGUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUAAAAUACACCACACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUGGUUAUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038110
miRBase ID: ami-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | aca-mir-99a |
Stem-loop Accession # | MI0018894 |
Stem-loop Sequence | GGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUAUAAUGCACAAGCUCGCUUCUAUGGGUCUGUGUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0021996
miRBase ID: aca-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | gga-mir-99a |
Stem-loop Accession # | MI0001173 |
Stem-loop Sequence | CCAAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGUUGAAAUGCACUGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUAUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0001103
miRBase ID: gga-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | cpi-mir-99a |
Stem-loop Accession # | MI0029444 |
Stem-loop Sequence | UGGUGCCAAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUAAAGUAUACUGCACAAGCUCGCUUCUAUGGGUCUGUGUCACUCUGGUGAUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0037710
miRBase ID: cpi-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | cli-mir-99 |
Stem-loop Accession # | MI0029945 |
Stem-loop Sequence | GAUGCCAAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGUUGAAACGCACUGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUAUGGUGAUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038478
miRBase ID: cli-miR-99-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | dno-mir-99a |
Stem-loop Accession # | MI0038954 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0047594
miRBase ID: dno-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | gmo-mir-99 |
Stem-loop Accession # | MI0036171 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGCAAAUAUGACAGCACAAGUUCGCUUCUGCAGGUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0044269
miRBase ID: gmo-miR-99-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | ggo-mir-99a |
Stem-loop Accession # | MI0002704 |
Stem-loop Sequence | CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002412
miRBase ID: ggo-miR-99a Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | cpo-mir-99a |
Stem-loop Accession # | MI0038610 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGUAAAGUGGACGGCACAAGCUCGCUUCUAUGGGUCUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0046941
miRBase ID: cpo-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | eca-mir-99a-2 |
Stem-loop Accession # | MI0012929 |
Stem-loop Sequence | CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCUCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0013184
miRBase ID: eca-miR-99a Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | ipu-mir-99a |
Stem-loop Accession # | MI0024668 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGCGUAAUCGGCAACCCAAGCUCGAUUCUGUGGGUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0029532
miRBase ID: ipu-miR-99a Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | lla-mir-99a |
Stem-loop Accession # | MI0002706 |
Stem-loop Sequence | CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002414
miRBase ID: lla-miR-99a Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | mne-mir-99a |
Stem-loop Accession # | MI0002707 |
Stem-loop Sequence | CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002415
miRBase ID: mne-miR-99a Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | mze-mir-99b |
Stem-loop Accession # | MI0034096 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGCGAAUCUGACAGCACAAGCUCGCCUCUGUGGGUCUU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042328
miRBase ID: mze-miR-99b Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | mmr-mir-99a |
Stem-loop Accession # | MI0039922 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGUAAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049202
miRBase ID: mmr-miR-99a Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | nbr-mir-99b |
Stem-loop Accession # | MI0034341 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGCAAAUGUGACAGCACAAGCUCGCCUCUGUGGGUCUU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042505
miRBase ID: nbr-miR-99b Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | oha-mir-99a |
Stem-loop Accession # | MI0031495 |
Stem-loop Sequence | GAGUGCCAAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAAUACUGUGCACAAGCUCGUUUCUAUGGGUCAGUGUCAGUCUGGUGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036964
miRBase ID: oha-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | oni-mir-99b |
Stem-loop Accession # | MI0034604 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGCAAAUCUGACAGCACAAGCUCGCCUCUGUGGGUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042695
miRBase ID: oni-miR-99b Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | ppa-mir-99a |
Stem-loop Accession # | MI0002708 |
Stem-loop Sequence | CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002416
miRBase ID: ppa-miR-99a Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | ptr-mir-99a |
Stem-loop Accession # | MI0002703 |
Stem-loop Sequence | CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002411
miRBase ID: ptr-miR-99a Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | ssc-mir-99a |
Stem-loop Accession # | MI0013114 |
Stem-loop Sequence | CCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0013896
miRBase ID: ssc-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | ppy-mir-99a |
Stem-loop Accession # | MI0002705 |
Stem-loop Sequence | CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUAAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002413
miRBase ID: ppy-miR-99a Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | pny-mir-99b |
Stem-loop Accession # | MI0034850 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGCGAAUCUGACAGCACAAGCUCGCCUCUGUGGGUCUU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042888
miRBase ID: pny-miR-99b Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | ocu-mir-99a |
Stem-loop Accession # | MI0039264 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0048182
miRBase ID: ocu-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | mml-mir-99a |
Stem-loop Accession # | MI0002702 |
Stem-loop Sequence | CCCAUUGGCAUAAACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGUCUGUGUCAGUGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002410
miRBase ID: mml-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | ssa-mir-99-1 |
Stem-loop Accession # | MI0026767 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGCAAAUCUGACAUCACAAGCUCCUCUCUGUGGGUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032718
miRBase ID: ssa-miR-99-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | ssa-mir-99-2 |
Stem-loop Accession # | MI0026768 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGCGAAUCUGACAUCACAAGCUCGCCUCUGUGGGCCUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032718
miRBase ID: ssa-miR-99-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | tch-mir-99a |
Stem-loop Accession # | MI0031223 |
Stem-loop Sequence | AACCCGUAGAUCCGAUCUUGUGGUGAAGUGGACCGCACAAGCUCGCUUCUAUGGGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036563
miRBase ID: tch-miR-99a-5p Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | xtr-mir-99 |
Stem-loop Accession # | MI0004821 |
Stem-loop Sequence | UAACAUAAACCCGUAGAUCCGAUCUUGUGGUGAAUUCCUUUGCUCAAGCUCGUUUCUAUGGGUCUGUGUCA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003582
miRBase ID: xtr-miR-99 Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | dre-mir-99-1 |
Stem-loop Accession # | MI0001956 |
Stem-loop Sequence | GCCACUUGUCAUUAACCCGUAGAUCCGAUCUUGUGAUAAGUUUGAUGGCACAAGCUCGAUUCUAUGGGUCUCUGUCUCUGUGGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0001812
miRBase ID: dre-miR-99 Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
Stem-loop ID | dre-mir-99-2 |
Stem-loop Accession # | MI0001957 |
Stem-loop Sequence | CACUUGUCACAAACCCGUAGAUCCGAUCUUGUGGCGUAAUCGGCAACCCAAGCUCGAUUCUGUGGGUCUCUGUCACUGUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0001812
miRBase ID: dre-miR-99 Mature miRNA Sequence: AACCCGUAGAUCCGAUCUUGUG Chromosome Location: NA |
More Information
Related Products
TaqMan™ Pri-miRNA Assay : Rn03465023_pri
TaqMan™ Pri-miRNA Assay : Mm03306206_pri
TaqMan™ Pri-miRNA Assay : Hs03302729_pri
Ambion® Pre-miR™ miRNA Precursor : PM10719
TaqMan™ MicroRNA Assay : 000435
mirVana® miRNA inhibitor : MH10719
mirVana® miRNA mimic : MC10719
TaqMan™ Advanced miRNA Assay : 478519_mir
TaqMan™ Advanced miRNA Assay : mmu478519_mir
TaqMan™ Advanced miRNA Assay : rno478519_mir
TaqMan™ Pri-miRNA Assay : Mm03306206_pri
TaqMan™ Pri-miRNA Assay : Hs03302729_pri
Ambion® Pre-miR™ miRNA Precursor : PM10719
TaqMan™ MicroRNA Assay : 000435
mirVana® miRNA inhibitor : MH10719
mirVana® miRNA mimic : MC10719
TaqMan™ Advanced miRNA Assay : 478519_mir
TaqMan™ Advanced miRNA Assay : mmu478519_mir
TaqMan™ Advanced miRNA Assay : rno478519_mir
miRBase Alias:
Human: hsa-miR-99a (v17)
Rat: rno-miR-99a (v18)
Aeromonas caviae Ae398: aca-miR-99a (v18)
Gallus gallus: gga-miR-99a (v18)
Macaca mulatta: mml-miR-99a (v19)
Macaca mulatta polyomavirus 1: mmu-miR-99a (v17)
Staphylococcus schleiferi: ssc-miR-99a (v21)
Rat: rno-miR-99a (v18)
Aeromonas caviae Ae398: aca-miR-99a (v18)
Gallus gallus: gga-miR-99a (v18)
Macaca mulatta: mml-miR-99a (v19)
Macaca mulatta polyomavirus 1: mmu-miR-99a (v17)
Staphylococcus schleiferi: ssc-miR-99a (v21)