Assay Details
Mature miRNA Sequence: | CUGCCACGAGCGUGCGGGCCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-12121 | MIMAT0049015 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-12121 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0039723 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.14: 104865314 - 104865411 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | CUGCCACGAGCGUGCGGGCCU | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | CUGCCACGAGCGUGCGGGCCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-12121 | MIMAT0049015 |
Stem-loop ID | hsa-mir-12121 |
Stem-loop Accession # | MI0039723 |
Stem-loop Sequence | UGGGCUCGGCCCGGGCUGCCACGAGCGUGCGGGCCUCGCCGGGCAUGUCCUAGGCGGCGGCCCCGCCCAGCGCUCGGCCGGGCGGGCGGGCGGGCGCG |
Chromosome Location | Chr. 14 - 104865314 - 104865411 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0049015
miRBase ID: hsa-miR-12121 Mature miRNA Sequence: CUGCCACGAGCGUGCGGGCCU Chromosome Location: Chr. 14 - 104865314 - 104865411 [+] on Build GRCh38 |