Assay Details
Mature miRNA Sequence: | UGGGCUCAGGGUACAAAGGUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4437 | MIMAT0018953 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4437 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0016778 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.2: 181305593 - 181305652 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UGGGCUCAGGGUACAAAGGUU | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UGGGCUCAGGGUACAAAGGUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4437 | MIMAT0018953 |
Stem-loop ID | hsa-mir-4437 |
Stem-loop Accession # | MI0016778 |
Stem-loop Sequence | ACUUUGUGCAUUGGGUCCACAAGGAGGGGAUGACCCUUGUGGGCUCAGGGUACAAAGGUU |
Chromosome Location | Chr. 2 - 181305593 - 181305652 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0018953
miRBase ID: hsa-miR-4437 Mature miRNA Sequence: UGGGCUCAGGGUACAAAGGUU Chromosome Location: Chr. 2 - 181305593 - 181305652 [-] on Build GRCh38 |