Assay Details
Gene Family ID: | MIPF0000615, mir-1266 |
Mature miRNA Sequence: | CCCUGUUCUAUGCCCUGAGGGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1266-3p | MIMAT0026742 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-1266-3p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0006403 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.15: 52277117 - 52277200 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | CCCUGUUCUAUGCCCUGAGGGA | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000615, mir-1266 |
Mature miRNA Sequence: | CCCUGUUCUAUGCCCUGAGGGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1266-3p | MIMAT0026742 |
Stem-loop ID | hsa-mir-1266 |
Stem-loop Accession # | MI0006403 |
Stem-loop Sequence | ACAGGUAGUGUCCCUCAGGGCUGUAGAACAGGGCUGGGAUUACUAAAGCCCUGUUCUAUGCCCUGAGGGACACUGAGCAUGUCA |
Chromosome Location | Chr. 15 - 52277117 - 52277200 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0026742
miRBase ID: hsa-miR-1266-3p Mature miRNA Sequence: CCCUGUUCUAUGCCCUGAGGGA Chromosome Location: Chr. 15 - 52277117 - 52277200 [-] on Build GRCh38 |