Assay Details
Mature miRNA Sequence: | UGGGGGUGUGGGGAGAGAGAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-7110-5p | MIMAT0028117 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-7110-5p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0022961 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.3: 123161794 - 123161879 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UGGGGGUGUGGGGAGAGAGAG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UGGGGGUGUGGGGAGAGAGAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-7110-5p | MIMAT0028117 |
Stem-loop ID | hsa-mir-7110 |
Stem-loop Accession # | MI0022961 |
Stem-loop Sequence | GGGGCUGGGGGUGUGGGGAGAGAGAGUGCACAGCCAGCUCAGGGAUUAAAGCUCUUUCUCUCUCUCUCUCUCCCACUUCCCUGCAG |
Chromosome Location | Chr. 3 - 123161794 - 123161879 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0028117
miRBase ID: hsa-miR-7110-5p Mature miRNA Sequence: UGGGGGUGUGGGGAGAGAGAG Chromosome Location: Chr. 3 - 123161794 - 123161879 [+] on Build GRCh38 |