Assay Details
Mature miRNA Sequence: | UUGCCUGCCCUCUUCCUCCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6894-3p | MIMAT0027689 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6894-3p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0022741 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.X: 53198889 - 53198945 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UUGCCUGCCCUCUUCCUCCAG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UUGCCUGCCCUCUUCCUCCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6894-3p | MIMAT0027689 |
Stem-loop ID | hsa-mir-6894 |
Stem-loop Accession # | MI0022741 |
Stem-loop Sequence | CAAGAAGGAGGAUGGAGAGCUGGGCCAGACAUGCUCUUGCCUGCCCUCUUCCUCCAG |
Chromosome Location | Chr. X - 53198889 - 53198945 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027689
miRBase ID: hsa-miR-6894-3p Mature miRNA Sequence: UUGCCUGCCCUCUUCCUCCAG Chromosome Location: Chr. X - 53198889 - 53198945 [-] on Build GRCh38 |