Assay Details
Mature miRNA Sequence: | ACUGGCAGAACACUGAAGCAGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6868-5p | MIMAT0027636 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6868-5p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0022715 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.17: 76098019 - 76098076 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | ACUGGCAGAACACUGAAGCAGC | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | ACUGGCAGAACACUGAAGCAGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6868-5p | MIMAT0027636 |
Stem-loop ID | hsa-mir-6868 |
Stem-loop Accession # | MI0022715 |
Stem-loop Sequence | CCAGGACUGGCAGAACACUGAAGCAGCAGGCACCUGCUUCCUUCUGUUGUCUGUGCAG |
Chromosome Location | Chr. 17 - 76098019 - 76098076 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027636
miRBase ID: hsa-miR-6868-5p Mature miRNA Sequence: ACUGGCAGAACACUGAAGCAGC Chromosome Location: Chr. 17 - 76098019 - 76098076 [-] on Build GRCh38 |