Assay Details
Mature miRNA Sequence: | UGACCUUUGCCUCUCCCCUCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6759-3p | MIMAT0027419 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6759-3p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0022604 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.12: 57748618 - 57748682 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UGACCUUUGCCUCUCCCCUCAG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UGACCUUUGCCUCUCCCCUCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6759-3p | MIMAT0027419 |
Stem-loop ID | hsa-mir-6759 |
Stem-loop Accession # | MI0022604 |
Stem-loop Sequence | UAUUGUUGUGGGUGGGCAGAAGUCUGUUUUCUUCAUGGUUUUCUGACCUUUGCCUCUCCCCUCAG |
Chromosome Location | Chr. 12 - 57748618 - 57748682 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027419
miRBase ID: hsa-miR-6759-3p Mature miRNA Sequence: UGACCUUUGCCUCUCCCCUCAG Chromosome Location: Chr. 12 - 57748618 - 57748682 [-] on Build GRCh38 |