Assay Details
Mature miRNA Sequence: | UCCUGCCUUCCUCUGCACCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6747-3p | MIMAT0027395 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6747-3p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0022592 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.11: 62567011 - 62567071 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UCCUGCCUUCCUCUGCACCAG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UCCUGCCUUCCUCUGCACCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6747-3p | MIMAT0027395 |
Stem-loop ID | hsa-mir-6747 |
Stem-loop Accession # | MI0022592 |
Stem-loop Sequence | UUUGGAGGGGUGUGGAAAGAGGCAGAACAUUCGUUCACUUUCCUGCCUUCCUCUGCACCAG |
Chromosome Location | Chr. 11 - 62567011 - 62567071 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027395
miRBase ID: hsa-miR-6747-3p Mature miRNA Sequence: UCCUGCCUUCCUCUGCACCAG Chromosome Location: Chr. 11 - 62567011 - 62567071 [-] on Build GRCh38 |