Assay Details
Mature miRNA Sequence: | CAGCAGGAGGUGAGGGGAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6165 | MIMAT0024782 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6165 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0021472 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.17: 49510817 - 49510900 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | CAGCAGGAGGUGAGGGGAG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | CAGCAGGAGGUGAGGGGAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6165 | MIMAT0024782 |
Stem-loop ID | hsa-mir-6165 |
Stem-loop Accession # | MI0021472 |
Stem-loop Sequence | CAGCAGGUCAGCAGGAGGUGAGGGGAGAGGAUCCACCUGUCCUGUCCUGUCCUCUCCUGCCCUGUCCUGGCUCCAGCCCCUCCC |
Chromosome Location | Chr. 17 - 49510817 - 49510900 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0024782
miRBase ID: hsa-miR-6165 Mature miRNA Sequence: CAGCAGGAGGUGAGGGGAG Chromosome Location: Chr. 17 - 49510817 - 49510900 [+] on Build GRCh38 |