Assay Details
Mature miRNA Sequence: | UCUGGGCACAGGCGGAUGGACAGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5189-5p | MIMAT0021120 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-5189-5p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0018168 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.16: 88468918 - 88469031 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UCUGGGCACAGGCGGAUGGACAGG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UCUGGGCACAGGCGGAUGGACAGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5189-5p | MIMAT0021120 |
Stem-loop ID | hsa-mir-5189 |
Stem-loop Accession # | MI0018168 |
Stem-loop Sequence | GGCCCGCCUUUUAGGGGCCUCGCUGUCUGGGCACAGGCGGAUGGACAGGCUGGCCUCUGGAUGACCUGCCAACCGUCAGAGCCCAGACCCACGUGGCCUCAGUUGGGGACCAGG |
Chromosome Location | Chr. 16 - 88468918 - 88469031 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0021120
miRBase ID: hsa-miR-5189-5p Mature miRNA Sequence: UCUGGGCACAGGCGGAUGGACAGG Chromosome Location: Chr. 16 - 88468918 - 88469031 [+] on Build GRCh38 |