Assay Details
Mature miRNA Sequence: | GAGGGCAUGCGCACUUUGUCC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4761-3p | MIMAT0019909 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4761-3p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0017402 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.22: 19963753 - 19963834 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | GAGGGCAUGCGCACUUUGUCC | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | GAGGGCAUGCGCACUUUGUCC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4761-3p | MIMAT0019909 |
Stem-loop ID | hsa-mir-4761 |
Stem-loop Accession # | MI0017402 |
Stem-loop Sequence | GGACAAGGUGUGCAUGCCUGACCCGUUGUCAGACCUGGAAAAAGGGCCGGCUGUGGGCAGGGAGGGCAUGCGCACUUUGUCC |
Chromosome Location | Chr. 22 - 19963753 - 19963834 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019909
miRBase ID: hsa-miR-4761-3p Mature miRNA Sequence: GAGGGCAUGCGCACUUUGUCC Chromosome Location: Chr. 22 - 19963753 - 19963834 [+] on Build GRCh38 |