Assay Details
Mature miRNA Sequence: | AAGUUGGCUGCAGUUAAGGUGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4715-5p | MIMAT0019824 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4715-5p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0017349 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.15: 25848747 - 25848825 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | AAGUUGGCUGCAGUUAAGGUGG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | AAGUUGGCUGCAGUUAAGGUGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4715-5p | MIMAT0019824 |
Stem-loop ID | hsa-mir-4715 |
Stem-loop Accession # | MI0017349 |
Stem-loop Sequence | GGGGAAUGAAAGUUGGCUGCAGUUAAGGUGGCUAAUCAGCUGAUGGUGCCACCUUAACUGCAGCCAAUUCUAAUUCCCC |
Chromosome Location | Chr. 15 - 25848747 - 25848825 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019824
miRBase ID: hsa-miR-4715-5p Mature miRNA Sequence: AAGUUGGCUGCAGUUAAGGUGG Chromosome Location: Chr. 15 - 25848747 - 25848825 [-] on Build GRCh38 |