Assay Details
Mature miRNA Sequence: | CUCUCUACUGACUUGCAACAUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4684-5p | MIMAT0019769 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4684-5p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0017316 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.1: 22719517 - 22719598 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | CUCUCUACUGACUUGCAACAUA | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | CUCUCUACUGACUUGCAACAUA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4684-5p | MIMAT0019769 |
Stem-loop ID | hsa-mir-4684 |
Stem-loop Accession # | MI0017316 |
Stem-loop Sequence | GCACCAGGGGUACCUCUCUACUGACUUGCAACAUACAUUUGUCUUGGUGUGUUGCAAGUCGGUGGAGACGUACCCUUGGUGC |
Chromosome Location | Chr. 1 - 22719517 - 22719598 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019769
miRBase ID: hsa-miR-4684-5p Mature miRNA Sequence: CUCUCUACUGACUUGCAACAUA Chromosome Location: Chr. 1 - 22719517 - 22719598 [+] on Build GRCh38 |