Assay Details
Mature miRNA Sequence: | GAAGAUGGUGCUGUGCUGAGGAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4647 | MIMAT0019709 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4647 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0017274 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.6: 44254206 - 44254285 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | GAAGAUGGUGCUGUGCUGAGGAA | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | GAAGAUGGUGCUGUGCUGAGGAA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4647 | MIMAT0019709 |
Stem-loop ID | hsa-mir-4647 |
Stem-loop Accession # | MI0017274 |
Stem-loop Sequence | CCAGGAGGGUGAAGAUGGUGCUGUGCUGAGGAAAGGGGAUGCAGAGCCCUGCCCAGCACCACCACCUCCUAUGCUCCUGG |
Chromosome Location | Chr. 6 - 44254206 - 44254285 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019709
miRBase ID: hsa-miR-4647 Mature miRNA Sequence: GAAGAUGGUGCUGUGCUGAGGAA Chromosome Location: Chr. 6 - 44254206 - 44254285 [-] on Build GRCh38 |