Assay Details
Mature miRNA Sequence: | UUAGUCCUGCCUGUAGGUUUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4540 | MIMAT0019083 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4540 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0016911 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.9: 36864254 - 36864308 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UUAGUCCUGCCUGUAGGUUUA | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UUAGUCCUGCCUGUAGGUUUA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4540 | MIMAT0019083 |
Stem-loop ID | hsa-mir-4540 |
Stem-loop Accession # | MI0016911 |
Stem-loop Sequence | AAGCUGCAUGGACCAGGACUUGGCACCUUUGGCCUUAGUCCUGCCUGUAGGUUUA |
Chromosome Location | Chr. 9 - 36864254 - 36864308 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019083
miRBase ID: hsa-miR-4540 Mature miRNA Sequence: UUAGUCCUGCCUGUAGGUUUA Chromosome Location: Chr. 9 - 36864254 - 36864308 [-] on Build GRCh38 |