Assay Details
Gene Family ID: | MIPF0001451, mir-4529 |
Mature miRNA Sequence: | AGGCCAUCAGCAGUCCAAUGAA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4529-5p | MIMAT0019236 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4529-5p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0016896 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.18: 55479221 - 55479298 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AGGCCAUCAGCAGUCCAAUGAA | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0001451, mir-4529 |
Mature miRNA Sequence: | AGGCCAUCAGCAGUCCAAUGAA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4529-5p | MIMAT0019236 |
Stem-loop ID | hsa-mir-4529 |
Stem-loop Accession # | MI0016896 |
Stem-loop Sequence | AUGACAGGCCAUCAGCAGUCCAAUGAAGACAUGAAGACCCAAUGUCUUCAUUGGACUGCUGAUGGCCCGUCACUGGGA |
Chromosome Location | Chr. 18 - 55479221 - 55479298 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019236
miRBase ID: hsa-miR-4529-5p Mature miRNA Sequence: AGGCCAUCAGCAGUCCAAUGAA Chromosome Location: Chr. 18 - 55479221 - 55479298 [+] on Build GRCh38 |