Assay Details
Mature miRNA Sequence: | AGACUGACGGCUGGAGGCCCAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4513 | MIMAT0019050 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4513 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0016879 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.15: 74788672 - 74788757 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | AGACUGACGGCUGGAGGCCCAU | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | AGACUGACGGCUGGAGGCCCAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4513 | MIMAT0019050 |
Stem-loop ID | hsa-mir-4513 |
Stem-loop Accession # | MI0016879 |
Stem-loop Sequence | AUUCUAGGUGGGGAGACUGACGGCUGGAGGCCCAUAAGCUGUCUAAAACUUCGGCCCCCAGAUUUCUGGUCUCCCCACUUCAGAAC |
Chromosome Location | Chr. 15 - 74788672 - 74788757 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019050
miRBase ID: hsa-miR-4513 Mature miRNA Sequence: AGACUGACGGCUGGAGGCCCAU Chromosome Location: Chr. 15 - 74788672 - 74788757 [-] on Build GRCh38 |